Merck
  • Mitochondrial very-long-chain acyl-coenzyme A dehydrogenase deficiency: clinical characteristics and diagnostic considerations in 30 patients.
Mitochondrial very-long-chain acyl-coenzyme A dehydrogenase deficiency: clinical characteristics and diagnostic considerations in 30 patients.
Clinica chimica acta; international journal of clinical chemistry (1998-03-14)
C Vianey-Saban, P Divry, M Brivet, M Nada, M T Zabot, M Mathieu, C Roe
ABSTRACT
Very-long-chain acyl-CoA dehydrogenase (VLCAD) is an enzyme catalyzing the dehydrogenation of long-chain fatty acids in the first step of mitochondrial fatty acid oxidation. Using an ETF (electron transfer flavoprotein, the physiological electron acceptor of VLCAD) reduction assay, we identified VLCAD deficiency in cultured skin fibroblasts or liver tissue from 30 patients in 27 families. They clinically presented two phenotypes: a 'severe' presentation characterized by an early onset of symptoms, with hypertrophic cardiomyopathy and a high incidence of death, and a 'mild' form with hypoketotic hypoglycaemia, resembling MCAD (medium-chain acyl-CoA dehydrogenase) deficiency. Cells isolated from patients who develop cardiomyopathy characteristically accumulate longer-chain length acylcarnitines (hexadecanoylcarnitine and tetradecanoylcarnitine) when incubated with palmitate. However, cells from patients with the hypoglycaemic presentation produced relatively shorter-chain-length intermediates (mainly dodecanoylcarnitine). Inhibition of carnitine palmitoyl transferase I, in vitro, eliminated these intermediates with cells from both phenotypes indicating their intramitochondrial origin. Although the explanation for these distinct biochemical findings is not obvious, the correlation with the two phenotypes provides an opportunity for accurate prognosis and early implementation of appropriate treatment. Prenatal diagnosis of this life-threatening disorder was successfully performed in seven pregnancies in six of those families by assay of trophoblasts or amniocytes. In an at risk family, diagnosis of an affected fetus by measurement of VLCAD activity in noncultured chorionic villi allowed termination of the pregnancy before 13 weeks of gestation.
Sigma-Aldrich
Stearoyl-L-carnitine, ≥97.0% (TLC)
Sigma-Aldrich
Lauroyl-L-carnitine, ≥95.0% (HPLC)