MilliporeSigma
Mutations in the kinase Rsk-2 associated with Coffin-Lowry syndrome.
Nature (1996-12-12)
E Trivier, D De Cesare, S Jacquot, S Pannetier, E Zackai, I Young, J L Mandel, P Sassone-Corsi, A Hanauer
ABSTRACT
The Coffin-Lowry syndrome (CLS), an X-linked disorder, is characterized by severe psychomotor retardation, facial and digital dysmorphisms, and progressive skeletal deformations. Genetic linkage analysis mapped the CLS locus to an interval of 2-3 megabases at Xp22.2. The gene coding for Rsk-2, a member of the growth-factor-regulated protein kinases, maps within the candidate interval, and was tested as a candidate gene for CLS. Initial screening for mutations in the gene for Rsk-2 in 76 unrelated CLS patients revealed one intragenic deletion, a nonsense, two splice site, and two missense mutations. The two missenses affect sites critical for the function of Rsk-2. The mutated Rsk-2 proteins were found to be inactive in a S6 kinase assay. These findings provide direct evidence that abnormalities in the MAPK/RSK signalling pathway cause Coffin-Lowry syndrome.