Merck
  • MIC2 is a specific marker for Ewing's sarcoma and peripheral primitive neuroectodermal tumors. Evidence for a common histogenesis of Ewing's sarcoma and peripheral primitive neuroectodermal tumors from MIC2 expression and specific chromosome aberration.
MIC2 is a specific marker for Ewing's sarcoma and peripheral primitive neuroectodermal tumors. Evidence for a common histogenesis of Ewing's sarcoma and peripheral primitive neuroectodermal tumors from MIC2 expression and specific chromosome aberration.
Cancer (1991-04-01)
I M Ambros, P F Ambros, S Strehl, H Kovar, H Gadner, M Salzer-Kuntschik
ABSTRACT
This study reports on the specific expression of the MIC2 gene, a pseudoautosomal gene located on the short arms of the X and Y chromosomes, on Ewing's sarcoma (ES) and peripheral primitive neuroectodermal tumor (pPNET) cells. The gene product, a cell membrane protein, is recognized by the newly established monoclonal antibody (MoAb) HBA-71 and the previously described MoAb 12E7 and RFB-1. Furthermore, the reaction pattern of the MIC2 antibodies, especially HBA-71, with normal tissues and a great number of benign and malignant tumors (70 different tumors, 199 tumor samples), as well as the correlation between the specific chromosomal aberrations, i.e., the t(11;22) and the del(22) and the expression of this antigen, are demonstrated. Both ES and pPNET cells express the MIC2 gene in very high amounts, which represents a highly selective and almost unique feature of these cells, making an assignment of these tumors in one entity even more likely. The MIC2 antibodies are of great value for clinical and research purposes.
Sigma-Aldrich
CD99 (EPR3097Y) Rabbit Monoclonal Primary Antibody