Merck
  • Serum vascular adhesion protein-1 predicts all-cause mortality and cancer-related mortality in subjects with colorectal cancer.
Serum vascular adhesion protein-1 predicts all-cause mortality and cancer-related mortality in subjects with colorectal cancer.
Clinica chimica acta; international journal of clinical chemistry (2013-11-12)
Yu-I Li, Ji-Shiang Hung, Tse-Ya Yu, Jyh-Ming Liou, Jung-Nan Wei, Hsien-Li Kao, Lee-Ming Chuang, Chia-Tung Shun, Po-Huang Lee, Hong-Shiee Lai, Chien-Yin Su, Hung-Yuan Li, Jin-Tung Liang
ABSTRACT
Vascular adhesion protein-1 (VAP-1) participates in inflammation and catalyzes the breakdown of amines to produce aldehyde, hydrogen peroxide, and ammonia. Serum VAP-1 can predict cancer mortality, including colorectal cancer (CRC) mortality, in type 2 diabetic subjects. However, it remains unknown if serum VAP-1 can predict mortality in CRC patients. This prospective cohort study investigates if serum VAP-1 is a novel biomarker for mortality prediction in CRC. We enrolled 300 CRC patients. Preoperative serum VAP-1 was measured by time-resolved immunofluorometric assay. They were followed until September 2009 or death, which was ascertained by the National Death Registration System. The median follow-up period was 4.7 years. Compared with normal counterpart, VAP-1 immunoactivity was upregulated in CRC tissues, especially at the invasion front. Serum VAP-1 can independently predict all-cause mortality (HR: 1.0026, 95% CI: 1.0003-1.0050, P<0.05) and cancer-related mortality (HR: 1.0026, 95% CI: 1.0001-1.0050, P<0.05). A risk score composed of age, gender, carcinoembryonic antigen (CEA) >5 ng/ml, tumor grading, tumor staging, and serum VAP-1 could stratify CRC patients into low-, intermediate-, and high-risk subgroups, with a 5-year mortality rate of 10%, 34%, and 78%, respectively. Serum VAP-1 predicts mortality independently and improves risk stratification in CRC subjects.