MilliporeSigma
  • Polymeric ligands as homogeneous, reusable catalyst systems for copper assisted click chemistry.
Polymeric ligands as homogeneous, reusable catalyst systems for copper assisted click chemistry.
Chemical communications (Cambridge, England) (2010-08-21)
Mieke Lammens, Jared Skey, Sofie Wallyn, Rachel O'Reilly, Filip Du Prez
ABSTRACT
Tris-(benzyltriazolylmethyl)amine (TBTA) has been immobilized onto a styrenic monomer and subsequently copolymerized with styrene to afford catalytically active and reusable copolymers for the CuAAC reaction.