Merck
  • Spectrophotometric study of Cd(II), Pb(II), Hg(II) and Zn(II) complexes with 5,10,15,20-tetrakis(4-carboxylphenyl)porphyrin.
Spectrophotometric study of Cd(II), Pb(II), Hg(II) and Zn(II) complexes with 5,10,15,20-tetrakis(4-carboxylphenyl)porphyrin.
Talanta (2008-10-31)
Krzysztof Kilian, Krystyna Pyrzyńska
ABSTRACT
The reaction of 5,10,15,20-tetrakis(4-carboxylphenyl)porphyrin (TCPP) with Cd(II), Pb(II), Hg(II) and Zn(II) was studied spectrophotometrically and kinetics, equilibrium constants as well as photodecomposition of complexes were determined. It was verified that these metal ions with large radius accelerate the incorporation reaction of zinc into TCPP. On the basis of the mechanism and kinetics of this reaction, a sensitive method for the spectrophotometric determination of trace amounts of Zn(II) has been developed. The molar absorptivity of examined Zn-TCPP complex and Sandell's sensitivity at 423 nm were 3.5x10(5) M(-1) cm(-1) and 18.3 ng cm(-2). The detection limit for the recommended procedure was 1.4x10(-9) M (0.9 ng ml(-1)) and precision in range 20-100 ng ml(-1) not exceeds 2.7% RSD. The proposed method applied for zinc determination in natural waters and nutritional supplement was compared with AAS results and declared value.
Sigma-Aldrich
Coproporphyrin I dihydrochloride, synthetic
Pricing and availability is not currently available.