Merck
  • Fibulin-2 exhibits high degree of variability, but no structural changes concordant with abdominal aortic aneurysms.
Fibulin-2 exhibits high degree of variability, but no structural changes concordant with abdominal aortic aneurysms.
European journal of human genetics : EJHG (1999-01-15)
H Kuivaniemi, A Marshall, A Ganguly, M L Chu, W M Abbott, G Tromp
ABSTRACT
We used conformation sensitive gel electrophoresis and direct sequencing of PCR products to screen for mutations in the cDNA for fibulin-2, an extracellular matrix protein, from 11 patients with abdominal aortic aneurysms and two controls. When compared with the published reference sequence, a total of 14 single-base sequence variations were detected. Seven of the changes were neutral in that they did not result in an amino acid substitution. There were five missense changes at sites not conserved between human and mouse, and two missense changes at sites conserved between human and mouse. All but two of the sequence variants studied were also present in an additional set of 102 control alleles analyzed. One of these two changes was a missense mutation, but it did not segregate with abdominal aortic aneurysms in the family, whilst the other change was neutral. In conclusion, fibulin-2 has a large number of sequence variations in comparison with our previous analyses of type III collagen, and these variations will be useful in association studies. There was an excellent overall agreement between direct sequencing of PCR-products and conformation sensitive gel electrophoresis.