Merck
  • Synthesis of 5-Substituted Derivatives of Isophthalic Acid as Non-Polymeric Amphiphilic Coating for Metal Oxide Nanoparticles.
Synthesis of 5-Substituted Derivatives of Isophthalic Acid as Non-Polymeric Amphiphilic Coating for Metal Oxide Nanoparticles.
Tetrahedron letters (2014-08-26)
Denis Nilov, Pavel Kucheryavy, Verina Walker, Clayton Kidd, Vladimir L Kolesnichenko, Galina Z Goloverda
ABSTRACT
In the course of development of novel capping ligands with variable steric factor, which will be used as an organic coating for metal oxide nanoparticles, a base-catalyzed nucleophilic oxirane ring-opening addition reaction between dimethyl 5-hydroxyisophthalate and allyl glycidyl ether was studied. The allyl-terminated 1-1, 1-2 and 1-3 adducts and dihydroxylated derivative of the 1-1 adduct, 5-diglyceroxy isophthalic acid, were synthesized. The latter binds to the surface of 5 nm γ-Fe
Sigma-Aldrich
Dimethyl 5-hydroxyisophthalate, 98%