Merck
  • End-binding protein 1 (EB1) up-regulation is an early event in colorectal carcinogenesis.
End-binding protein 1 (EB1) up-regulation is an early event in colorectal carcinogenesis.
FEBS letters (2014-02-05)
Yolanda Stypula-Cyrus, Nikhil N Mutyal, Mart Dela Cruz, Dhananjay P Kunte, Andrew J Radosevich, Ramesh Wali, Hemant K Roy, Vadim Backman
ABSTRACT
End-binding protein (EB1) is a microtubule protein that binds to the tumor suppressor adenomatous polyposis coli (APC). While EB1 is implicated as a potential oncogene, its role in cancer progression is unknown. Therefore, we analyzed EB1/APC expression at the earliest stages of colorectal carcinogenesis and in the uninvolved mucosa ("field effect") of human and animal tissue. We also performed siRNA-knockdown in colon cancer cell lines. EB1 is up-regulated in early and field carcinogenesis in the colon, and the cellular/nano-architectural effect of EB1 knockdown depended on the genetic context. Thus, dysregulation of EB1 is an important early event in colon carcinogenesis.