Merck
  • Prognostic value of centromere protein-A expression in patients with epithelial ovarian cancer.
Prognostic value of centromere protein-A expression in patients with epithelial ovarian cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2013-05-29)
Jun-Jun Qiu, Jun-Jie Guo, Tian-Jiao Lv, Hong-Yan Jin, Jing-Xin Ding, Wei-Wei Feng, Ying Zhang, Ke-Qin Hua
ABSTRACT
Altered expression of centromere protein-A (CENP-A) is observed in various types of human cancers. However, the clinical significance and pathological role of CENP-A in epithelial ovarian cancer (EOC) remains unclear. The main objective of this investigation was to clarify the relationships between CENP-A expression and the clinicopathological features of patients with EOC. Real-time quantitative PCR and Western blot were performed to examine CENP-A expression in 20 pairs of fresh-frozen EOC tissues and corresponding noncancerous tissues. Using immunohistochemistry, we performed a retrospective study of the CENP-A expression levels on 120 archival EOC paraffin-embedded samples. Prognostic outcomes correlated with CENP-A were examined using Kaplan-Meier analysis and Cox proportional hazards model. Our results showed that the expression levels of CENP-A mRNA and protein in EOC tissues were both significantly higher than those in noncancerous tissues. By immunohistochemistry, the data revealed that high CENP-A expression was significantly correlated with pathological grade (P = 0.02) and International Federation of Gynecology and Obstetrics stage (P = 0.006). Consistent with these results, we found that high expression of CENP-A was significantly correlated with poor survival in EOC patients (P < 0.001). Furthermore, Cox regression analyses showed that CENP-A expression was an independent predictor of overall survival. Our data suggest that CENP-A could play an important role in EOC and might serve as a valuable prognostic marker and potential target for gene therapy in the treatment of EOC.