Merck
Fusidic acid in dermatology: an updated review.
European journal of dermatology : EJD (2009-12-17)
Helmut Schöfer, Lene Simonsen
ABSTRACT
Studies on the clinical efficacy of fusidic acid in skin and soft-tissue infections (SSTIs), notably those due to Staphylococcus aureus, are reviewed. Oral fusidic acid (tablets dosed at 250 mg twice daily, or a suspension for paediatric use at 20 mg/kg/day given as two daily doses) has shown good efficacy and tolerability. Similarly, plain fusidic acid cream or ointment used two or three times daily in SSTIs such as impetigo are clinically and bacteriologically effective, with minimal adverse events. Combination formulations of fusidic acid with 1% hydrocortisone or 0.1% betamethasone achieve excellent results in infected eczema by addressing both inflammation and infection. A new lipid-rich combination formulation provides an extra moisturizing effect. Development of resistance to fusidic acid has remained generally low or short-lived and can be minimized by restricting therapy to no more than 14 days at a time.
Sigma-Aldrich
Fusidic acid
Fusidic acid, European Pharmacopoeia (EP) Reference Standard