Merck
  • Asymmetric printing of molecules and zeolites on self assembled monolayers.
Asymmetric printing of molecules and zeolites on self assembled monolayers.
Nanoscale (2010-07-21)
Nermin Seda Kehr, Andreas Schäfer, Bart Jan Ravoo, Luisa De Cola
ABSTRACT
Microcontact printing (mCP) is used to immobilize dyes and peptides asymmetrically, by a "peptide coupling" reaction, on monolayers of zeolite L crystals in the contact area between the stamp and the surface of the monolayer. Chemically patterned surfaces of monolayers of zeolite L crystals are obtained by using patterned stamps with different ink solutions. Additional printing of functionalized nano-objects on SAMs of zeolite L crystals is demonstrated.
Sigma-Aldrich
Atto 425, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425-Biotin, BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Sigma-Aldrich
Atto 425 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)