Merck
  • Synthesis of (S)-(+)-decursin and its analogues as potent inhibitors of melanin formation in B16 murine melanoma cells.
Synthesis of (S)-(+)-decursin and its analogues as potent inhibitors of melanin formation in B16 murine melanoma cells.
European journal of medicinal chemistry (2010-10-05)
Kyeong Lee, Jee-Hyun Lee, Shanthaveerappa K Boovanahalli, Yongseok Choi, Soo-Jin Choo, Ick-dong Yoo, Dong Hee Kim, Mi Young Yun, Gye Won Lee, Gyu-Yong Song
ABSTRACT
We report the synthesis of a novel series of highly potent melanin inhibitors which were obtained through structural modification of an anticancer compound S-(+)-decursinol. The in vitro inhibitory potencies of the newly synthesized compounds were evaluated against α-MSH induced melanin production in B16 murine melanoma cells. Among the compounds evaluated, compounds 2, 3, 6b, 7a, 7b, 8a and 8b emerged as highly potent inhibitors of melanin production. Besides, these compounds demonstrated significantly low cytotoxicity.
Sigma-Aldrich
Decursin, ≥97% (HPLC)