Merck
  • Determination of carbamazepine by chemiluminescence detection using chemically prepared tris(2,2'-bipyridine)ruthenium(III) as oxidant.
Determination of carbamazepine by chemiluminescence detection using chemically prepared tris(2,2'-bipyridine)ruthenium(III) as oxidant.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2003-07-02)
Sang Hak Lee, Ming Li, Jung Kee Suh
ABSTRACT
A new chemiluminescence method for the determination of carbamazepine (CBZ) has been developed. The method is based on the chemiluminescence produced in the reaction of tris(2,2'-bipyridine)ruthenium(III) and CBZ in an acidic medium. The chemiluminescence intensity was enhanced by organic solvents in the reaction system. Under the optimum experimental conditions, the calibration curve was linear over the range 4.0 x 10(-3)-8.6 x 10(-7) mol/L for CBZ. The detection limit (S/N = 3) was 2.5 x 10(-7) mol/L and the relative standard deviation of six replicate measurements was 2.6% for 4.0 x 10(-4) mol/L of CBZ. The possible reaction mechanism were also discussed. The chemiluminescence method was successfully applied to assay the CBZ contents in pharmaceutical tablets.
Sigma-Aldrich
10,11-Dihydrocarbamazepine, 99%