Merck
  • Association between thyroid profile and perfluoroalkyl acids: data from NHNAES 2007-2008.
Association between thyroid profile and perfluoroalkyl acids: data from NHNAES 2007-2008.
Environmental research (2013-09-24)
Ram B Jain
ABSTRACT
The effect of six perfluoroalkyl acids (PFAAs), namely, perfluorooctane sulfonate (PFOS), perfluorooctanoic acid (PFOA), perfluorodecanoic acid (PFDE), perfluorohexane sulfonic acid (PFHxS), 2-(N-methyl-perfluorooctane sulfonamide) acetic acid (MPAH), and perfluorononanoic acid (PFNA) on the levels of six thyroid function variables, namely, thyroid stimulating hormone (TSH), free and total thyroxine (FT4, TT4), free and total triiodothyronine (FT3, TT3), and thyroglobulin (TGN) was evaluated. Data from National Health and Nutrition Examination Survey for the years 2007-2008 were used for this evaluation. TSH levels increased with increase in levels of PFOA (p<0.01). There were no statistically significant associations between the levels of FT3, and FT4 with the levels of any of the six PFAAs. Levels of TT3 were found to increase with the levels of PFOA (p=0.01) and TT4 levels were found to increase with increase in PFHxS levels (p<0.01). Males had statistically significantly higher levels of FT3 than females and females had statistically significantly higher levels of TT4 than males. As compared to non-Hispanics whites and Hispanics, non-Hispanic blacks had lower levels of TSH, FT3, TT3, and TT4 but Hispanics had the lowest levels of TGN. Age was negatively associated with FT3 and TT3 but positively associated with FT4 and TT4. Non-smokers had higher levels of TSH and TT4 than smokers and smokers had higher levels of FT3 and TGN than non-smokers. Iodine deficiency was associated with increased levels of TSH, TT3, TT4, and TGN.
Sigma-Aldrich
Perfluorodecanoic acid, 97%