- Design and synthesis of potent N1-substituted indole melatonin receptor agonists.
Design and synthesis of potent N1-substituted indole melatonin receptor agonists.
Chemical communications (Cambridge, England) (2003-03-05)
Andrew Tsotinis, Andreas Eleutheriades, Katherine Hough, David Sugden
PMID12613623
ABSTRACT
The design and expeditious synthesis of two new indole analogs with up to 5-fold potency of that of melatonin is described.