- Gene expression network analysis of ETV1 reveals KCTD10 as a novel prognostic biomarker in gastrointestinal stromal tumor.
Gene expression network analysis of ETV1 reveals KCTD10 as a novel prognostic biomarker in gastrointestinal stromal tumor.
Prognostic biomarkers are required for risk stratification therapy in the patients with gastrointestinal stromal tumor (GIST). In this study, we aimed to identify prognostic biomarkers in GIST. We assessed the prognostic value of E twenty-six variant 1 (ETV1), a recently identified transcription factor unique to GIST. We also examined the clinical utility and functions of its downstream gene, potassium channel tetramerization domain containing protein 10 (KCTD10). The levels of ETV1 and KCTD10 were evaluated immunohistochemically in 112 patients with GIST treated at two hospitals. The functional properties of KCTD10 were examined by gene silencing assay in cultured GIST cells. Immunohistochemistry revealed that ETV1 expression in GIST had no prognostic significance. In contrast, the disease-free survival rate was 88.5% in patients with KCTD10-positive tumors and 55.8% in those with KCTD10-negative tumors (p <0.0001). KCTD10 was an independent prognostic factor (p <0.05). In the low-risk classification group, KCTD10 was significantly associated with favorable prognosis (p = 0.0008). Gene silencing of KCTD10 increased cell proliferation and invasion, suggesting that KCTD10 has a tumor-suppressive function. The GIST-specific transcription factor ETV1 may have no prognostic potential, whereas its downstream gene KCTD10 is associated with a favorable prognosis. Our study indicated the novel prognostic utility of KCTD10 in GIST, and suggested its tumor-suppressive effects on GIST cells. Further validation studies of KCTD10 for clinical applications, and functional verification of KCTD10 for better understanding of molecular basis of malignant phenotypes are worth challenging in GIST.