Merck
  • Triazole- and triazolium-containing porphyrin-cages for optical anion sensing.
Triazole- and triazolium-containing porphyrin-cages for optical anion sensing.
Dalton transactions (Cambridge, England : 2003) (2012-05-09)
Lydia C Gilday, Nicholas G White, Paul D Beer
ABSTRACT
Triazole and triazolium groups have been integrated into a zinc(II) metalloporphyrin-based structural framework to produce two porphyrin-cages for anion sensing applications. UV/visible spectroscopic titration investigations reveal both host systems exhibit strong anion binding affinities, with the positively-charged triazolium-porphyrin cage capable of colorimetric sensing halides, fluoride and chloride, and oxoanions in acetone-water solvent mixtures.
Sigma-Aldrich
3-(Bromomethyl)benzonitrile, 95%