Merck
  • Alkyl and carboxylalkyl esters of 4'-demethyl-4-deoxypodophyllotoxin: synthesis, cytotoxic, and antitumor activity.
Alkyl and carboxylalkyl esters of 4'-demethyl-4-deoxypodophyllotoxin: synthesis, cytotoxic, and antitumor activity.
European journal of medicinal chemistry (2004-02-28)
Young-Jae You, Yong Kim, Nguyen-Hai Nam, Seong-Cheol Bang, Byung-Zun Ahn
ABSTRACT
Esters of 4'-demethyl-4-deoxypodophyllotoxin (DDPT) with alkanoic acids and alkanedioic acids were prepared and tested for cytotoxic and antitumor activity. Among 19 esters, esters of propanoic acid, tetradecanedioic acid, 13-carboxyundecanoic acid, and hexadecanedioic acid improved the antitumor activity compared with that of the starting compounds, DDPT.
Sigma-Aldrich
Hexadecanedioic acid, 96%