Merck
  • Reversion of malignant phenotype by 5-iodo-6-amino-1,2-benzopyrone a non-covalently binding ligand of poly(ADP-ribose) polymerase.
Reversion of malignant phenotype by 5-iodo-6-amino-1,2-benzopyrone a non-covalently binding ligand of poly(ADP-ribose) polymerase.
Biochimie (1995-01-01)
P I Bauer, E Kirsten, G Varadi, L J Young, A Hakam, J A Comstock, E Kun
ABSTRACT
A non-covalently binding inhibitory ligand of poly(ADP-ribose) polymerase, 5-iodo-6-amino-1,2-benzopyrone, when incubated at 5-600 microM external concentration with an E-ras-transformed tumorigenic cell line or with human prostatic carcinoma cells for 40 to 60 days converts both cancer cells to a non-tumorigenic phenotype that is characterized by drastic changes in cell morphology, absence of tumorigenicity in nude mice, and a high rate of aerobic glycolysis.