Merck
  • Protection by phosphodiesterase inhibitors against endotoxin-induced liver injury in galactosamine-sensitized mice.
Protection by phosphodiesterase inhibitors against endotoxin-induced liver injury in galactosamine-sensitized mice.
Biochemical pharmacology (1993-06-22)
W Fischer, C Schudt, A Wendel
ABSTRACT
Phosphodiesterase inhibitors were used as a tool to manipulate cellular nucleotide levels in vitro and in vivo. The lipopolysaccharide (LPS)-induced release of tumor necrosis factor alpha (TNF-alpha) from mouse peritoneal macrophages was inhibited by prostaglandin E2 with an IC50 of 0.05 microM and by dibutyryl-cAMP with an IC50 of 180 microM. In the presence of the phosphodiesterase inhibitors zardaverine or rolipram the intracellular cAMP concentration of LPS-stimulated macrophages was significantly increased. In these cells, LPS-inducible TNF release was inhibited by zardaverine (IC50 = 1.5 microM) or by rolipram (IC50 = 0.35 microM). In a model of septic shock, i.e. LPS challenge of galactosamine-sensitized mice, a dose-dependent protection against liver injury was observed following oral application of rolipram (ED50 = 0.55 mg/kg) or of zardaverine (ED50 approximately 30 mg/kg). The adenylate cyclase activator forskolin was also protective. Rolipram also protected against TNF-induced liver injury in mice while zardaverine failed to do so. It is concluded that the intracellular cAMP level of macrophages is a critical determinant of LPS-inducible TNF release and therefore modulates the susceptibility to septic shock.
Sigma-Aldrich
Zardaverine