Merck
  • The immunosuppressive substance 2-chloro-2-deoxyadenosine modulates lipoprotein metabolism in a murine macrophage cell line (P388 cells).
The immunosuppressive substance 2-chloro-2-deoxyadenosine modulates lipoprotein metabolism in a murine macrophage cell line (P388 cells).
Lipids (1994-09-01)
M Lechleitner, B Auer, U Zilian, F Hoppichler, M Schirmer, B Föger, F Geisen, J R Patsch, G Konwalinka
ABSTRACT
A recently developed immunosuppressive substance, 2-chloro-2-deoxyadenosine (2-CdA), was reported to inhibit monocyte functions at low concentration. Because macrophages play a key role in the formation of atherosclerotic plaques, it was of interest to study the effect of 2-CdA on cellular lipid metabolism. For this purpose we have used a macrophage cell line (P388) to perform incubation studies in the presence of acetylated low density lipoprotein (Ac-LDL) and 2-CdA. The addition of 2-CdA, in concentrations ranging from 5-20 nM, induced a dose-dependent decrease in cellular cholesterol content and in the amount of extracellular [14C]oleic acid (OA) incorporated into the cholesteryl ester (CE) fraction. The effect was maximized at 20 nM 2-CdA with an 86% reduction in cholesterol esterification compared to controls (P < 0.008). To evaluate the mechanism of interaction of 2-CdA with cellular lipid metabolism, deoxycytidine (dCyt) and 3-methoxybenzamide (3-MOB), substances known to antagonize the effect of 2-CdA in different ways, were co-administered with 2-CdA. dCyt, a competitive inhibitor of dCyt kinase, which catalyzes phosphorylation to the active metabolite, antagonized the effects of 20 nM 2-CdA, producing significantly greater incorporation of extracellular [14C]OA into the CE fraction than in the presence of 2-CdA alone (P < 0.0086). Co-incubation with 2-CdA and the poly-ADP-ribose synthetase inhibitor 3-MOB, which is known to render cells resistant to 2-CdA toxicity by preventing cellular nicotinamide adenine dinucleotide (NAD)- and adenosine triphosphase-depletion, also reversed the effect of 2-CdA on lipid accumulation.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
3-Methoxybenzamide, 97%