Merck
  • beta 2-Inhibin contains the active core of human seminal plasma beta-inhibin: synthesis and bioactivity.
beta 2-Inhibin contains the active core of human seminal plasma beta-inhibin: synthesis and bioactivity.
FEBS letters (1985-02-11)
N J Arbatti, N G Seidah, J Rochemont, E Escher, A R Sheth, M Chrétien
ABSTRACT
The complete synthesis of the C-terminal 28 residues segment 67-94 of human seminal plasma beta-Inhibin, called beta 2-Inhibin, is reported. The Inhibin-like activity of the native 94 amino acids beta-Inhibin is compared to that of the synthetic replica of beta 2-Inhibin. In all assays used both peptides effectively suppress the FSH release induced by LHRH but have little effect on the LH release. In the mouse both peptides are equipotent on a mole basis. In the rat the synthetic beta 2-Inhibin is 3-10 times more potent than beta-Inhibin. Both peptides are active in rat anterior pituitary primary culture assays where maximum suppression of FSH release induced by LHRH occurs around 300 pmol/ml of beta 2-Inhibin. In contrast, maximum suppression of FSH release in the mouse pituitary assay occurs at 10-15 pmol/ml of either Inhibin.