Merck
  • Reductive dechlorination of hexachlorobenzene to tri- and dichlorobenzenes in anaerobic sewage sludge.
Reductive dechlorination of hexachlorobenzene to tri- and dichlorobenzenes in anaerobic sewage sludge.
Applied and environmental microbiology (1988-02-01)
B Z Fathepure, J M Tiedje, S A Boyd
ABSTRACT
Hexachlorobenzene was dechlorinated to tri- and dichlorobenzenes in anaerobic sewage sludge. The complete biotransformation of 190 microM hexachlorobenzene (approximately 50 ppm) occurred within 3 weeks. The calculated rate of hexachlorobenzene dechlorination was 13.6 mumol liter-1 day-1. Hexachlorobenzene was dechlorinated via two routes, both involving the sequential removal of chlorine from the aromatic ring. The major route was hexachlorobenzene----pentachlorobenzene----1,2,3,5-tetrachlorobenzene--- -1,3,5- trichlorobenzene. Greater than 90% of the added hexachlorobenzene was recovered as 1,3,5-trichlorobenzene, and there was no evidence for further dechlorination of 1,3,5-trichlorobenzene. The minor route was hexachlorobenzene----pentachlorobenzene----1,2,4,5-tetrachlorobenzene--- -1,2,4- trichlorobenzene----dichlorobenzenes. These results extend reductive dechlorination to poorly water soluble aromatic hydrocarbons which could potentially include other important environmental pollutants like polychlorinated biphenyls.
Product Description
Supelco
1,2,3,5-Tetrachlorobenzene, PESTANAL®, analytical standard