Merck
  • 1-O-hexadecyl-2-Q-methylglycerol, a novel inhibitor of protein kinase C, inhibits the respiratory burst in human neutrophils.
1-O-hexadecyl-2-Q-methylglycerol, a novel inhibitor of protein kinase C, inhibits the respiratory burst in human neutrophils.
The Journal of biological chemistry (1989-04-05)
I M Kramer, R L van der Bend, A T Tool, W J van Blitterswijk, D Roos, A J Verhoeven
ABSTRACT
To assess the role of protein kinase C (Ca2+/phospholipid-dependent enzyme) in the activation of the human neutrophil respiratory burst, we have utilized an ether lipid of the type 1-O-alkyl-2-O-methylglycerol (AMG), recently shown to be an inhibitor of this kinase. AMG-C16 (with an hexadecyl chain at the sn-1 position) was found to inhibit the respiratory burst induced by sub-optimal concentrations of phorbol 12,13-dibutyrate. Respiratory burst activity was recovered by subsequent addition of a supraoptimal dose of phorbol 12-myristate 13-acetate, indicating that in the presence of the inhibitor only the activation of the NADPH:O2 oxidoreductase via protein kinase C is inhibited, but not the oxidoreductase itself. The respiratory burst induced by the chemoattractant N-formyl-methionyl-leucyl-phenylalanine (fMLP) was also inhibited in the presence of AMG-C16, the extent of inhibition being dependent on the concentration of fMLP. At the concentrations applied in these studies, AMG-C16 had no effect on cell viability, did not affect the formation of inositol phosphates induced by fMLP, and did not affect the characteristics of the Ca2+ fluxes induced by the same stimulus. In a cell-free assay system, AMG-C16 had no effect on the activity of cAMP-dependent or Ca2+/calmodulin-dependent protein kinase but inhibited protein kinase C in a dose-dependent fashion. To characterize the inhibitory action of AMG-C16 on the respiratory burst activity in more detail, we studied protein phosphorylation in relation to respiratory burst activity in neutrophil cytoplasts. We focused on the phosphorylation of the 47-kDa protein, because this protein is functionally associated with the NADPH:O2 oxidoreductase. At suboptimal concentrations of phorbol 12,13-dibutyrate, AMG-C16 inhibited phosphorylation of proteins, including that of the 47-kDa protein. Recovery of protein phosphorylation in parallel to recovery of respiratory burst activity was obtained by addition of increasing doses of phorbol 12,13-dibutyrate. Recovery of respiratory burst activity at intermediate concentrations of fMLP did not result in a proportional increase in 47-kDa protein phosphorylation; phosphorylation of the 47-kDa protein was recovered only at high concentrations of fMLP. From these data we conclude that protein kinase C is involved in the activation of the respiratory burst by phorbol esters and fMLP. However, with fMLP as a stimulus, a second signal seems to be triggered, which is insensitive to AMG-C16.