Merck
  • Molecular recognition of floral volatile with two olfactory related proteins in the Eastern honeybee (Apis cerana).
Molecular recognition of floral volatile with two olfactory related proteins in the Eastern honeybee (Apis cerana).
International journal of biological macromolecules (2013-02-14)
Hongliang Li, Linya Zhang, Cuixia Ni, Hanwu Shang, Shulin Zhuang, Jianke Li
ABSTRACT
The honeybee relies on its sensitive olfaction to perform the foraging activities in the field. In the antennal chemoreception system of honeybee, odorant-binding proteins (OBPs) and chemosensory protein (CSPs) are major two protein families capable of binding with some plant volatiles and chemical ligands. However, the chemical binding interaction of plant odors with OBPs and CSPs in the honeybee olfactory system is still not clear yet. Hence, complex fluorescent spectra, ultraviolet absorption spectra, circular dichroism spectra and molecular docking were used to investigate the binding property of AcerASP2 (an OBP of Apis cerana) and AcerASP3 (a CSP of Apis cerana) with β-ionone, one of ordinary floral volatiles in botanical flower. As a result, β-ionone had a strong capability to quench the fluorescence that the two proteins produced, and their interaction was a dynamic process that was mainly driven by a hydrophobic force. AcerASP2 had a larger hydrophobic cavity than that of AcerASP3 and the conformation of AcerASP2 was changed less than AcerASP3 when binding with β-ionone. Our data suggests that OBPs like AcerASP2 might make a large contribution toward assisting the honeybee in sensing and foraging flowers, and A. cerana has evolved a good circadian rhythm to perceive a flower's odor following the fluctuation of temperature in the olfactory system. This significantly extends our knowledge on how to strengthen the honeybees' pollination service via manipulation of target proteins in the olfactory system.
Sigma-Aldrich
β-Ionone, natural, ≥95%, FG
Sigma-Aldrich
β-Ionone, 96%
Sigma-Aldrich
β-Ionone, predominantly trans, ≥97%, FCC, FG