- Influence of emotional stress on pharmacokinetics of isosorbide dinitrate intraperitoneally administered to rats.
Influence of emotional stress on pharmacokinetics of isosorbide dinitrate intraperitoneally administered to rats.
Acta medica Okayama (1990-02-01)
Y Gomita, K Furuno, Y Araki
PMID2330846
ABSTRACT
The plasma level of isosorbide dinitrate intraperitoneally administered to rats stressed by foot-shock was almost the same as that in non-stressed control rats. However, levels of its metabolites, 5-isosorbide mononitrate and 2-isosorbide mononitrate, were lower in stressed rats than in non-stressed rats, suggesting that stress may influence the metabolism and/or excretion of the metabolites.
Product Description
Isosorbide 2-nitrate, European Pharmacopoeia (EP) Reference Standard