Merck
  • Chemoenzymatic synthesis of cholesteryl-6-O-tetradecanoyl-α-D-glucopyranoside: a product of host cholesterol efflux promoted by Helicobacter pylori.
Chemoenzymatic synthesis of cholesteryl-6-O-tetradecanoyl-α-D-glucopyranoside: a product of host cholesterol efflux promoted by Helicobacter pylori.
Chemical communications (Cambridge, England) (2012-08-03)
Ryan A Davis, Chun-Hung Lin, Jacquelyn Gervay-Hague
ABSTRACT
In a three-step protocol involving regioselective enzymatic acylation, per-O-trimethylsilylation, and a one-pot α-glycosidation-deprotection sequence, cholesteryl-6-O-tetradecanoyl-α-D-glucopyranoside (α-CAG) of Helicobacter pylori is afforded starting from glucose in an overall yield of 45%. The production of CAG can be scaled to make purified quantities available to the biological community for the first time.
Sigma-Aldrich
Cholesterol β-D-glucoside, ≥97% (TLC)