Merck
Diverse trifluoromethyl heterocycles from a single precursor.
The Journal of organic chemistry (2012-01-24)
Mark A Honey, Raffaele Pasceri, William Lewis, Christopher J Moody
ABSTRACT
Ethyl 2-diazo-4,4,4-trifluoro-3-oxobutanoate is a highly versatile intermediate for the synthesis of a wide range of trifluoromethyl heterocycles. With the use of rhodium(II) or copper(II) catalyzed carbene X-H insertion reactions as key steps, a diverse set of trifluoromethyl-oxazoles, -thiazoles, -imidazoles, -1,2,4-triazines, and -pyridines are available from the diazoketoester, either directly in a single step or with just one additional step.