Merck
  • Dioxygenolytic cleavage of aryl ether bonds: 1,2-dihydro-1,2-dihydroxy-4-carboxybenzophenone as evidence for initial 1,2-dioxygenation in 3- and 4-carboxy biphenyl ether degradation.
Dioxygenolytic cleavage of aryl ether bonds: 1,2-dihydro-1,2-dihydroxy-4-carboxybenzophenone as evidence for initial 1,2-dioxygenation in 3- and 4-carboxy biphenyl ether degradation.
FEMS microbiology letters (1990-06-01)
K H Engesser, W Fietz, P Fischer, P Schulte, H J Knackmuss
ABSTRACT
A bacterial strain, Pseudomonas sp. POB 310, was enriched with 4-carboxy biphenyl ether as sole source of carbon and energy. Resting cells of POB 310 co-oxidize a substrate analogue, 4-carboxybenzophenone, yielding 1,2-dihydro-1,2-dihydroxy-4-carboxy-benzophenone. The ether bond of 3- and 4-carboxy biphenyl ether is cleaved analogously by initial 1,2-dioxygenation, yielding a hemiacetal which is hydrolysed to protocatechuate and phenol. These intermediates are degraded via an ortho and meta pathway, respectively. Alternative 2,3- and 3,4-dioxygenation can be ruled out as triggering steps in carboxy biphenyl ether degradation.
Sigma-Aldrich
4-Phenoxybenzoic acid, 97%