Merck
  • The risk of ischaemic colitis in irritable bowel syndrome patients treated with serotonergic therapies.
The risk of ischaemic colitis in irritable bowel syndrome patients treated with serotonergic therapies.
Drug safety (2011-06-15)
James H Lewis
ABSTRACT
Ischaemic colitis (IC) is the most common form of ischaemic injury to the gastrointestinal (GI) tract. IC typically presents with the sudden onset of lower abdominal pain, cramping and rectal bleeding, and is usually self-limited with low morbidity, although it may cause gangrenous or fulminant colitis, especially when the right colon is involved. Multiple medical conditions, as well as several pharmacological agents, are associated with IC, including irritable bowel syndrome (IBS) and drugs used for its treatment that act on gut serotonin 5-HT receptors. These include the selective 5-HT(3) receptor antagonist alosetron, currently approved for the treatment of severe diarrhoea-predominant IBS in women who fail to respond to conventional treatment, and cilansetron, another 5-HT(3) receptor antagonist that is no longer in clinical development. In addition, the 5-HT(4) receptor partial agonist tegaserod, which was approved for the treatment of constipation-predominant IBS in women, was associated with IC in the postmarketing setting, as was renzapride, a 5-HT(4) agonist/5-HT(3) antagonist. Although several hypotheses have been proposed, the pathophysiological basis for development of IC with 5-HT(3) receptor antagonists or 5-HT(4) receptor agonists remains unknown. Of interest, several population-based studies demonstrated that a diagnosis of IBS (independent of serotonergic therapies) increases the risk of developing IC 2- to 4-fold. As a result, IBS patients with the acute onset of abdominal pain, tenderness, diarrhoea or lower intestinal bleeding, especially those with predisposing conditions or medications, should be evaluated promptly for IC. The management of IC remains supportive; most cases of non-gangrenous IC, as seen in the alosetron and tegaserod databases, have been transient and have resolved spontaneously without complications or death. Despite the small number of deaths associated with alosetron in patients with complications of constipation and because of the ongoing requirement to prescribe alosetron under a risk management plan, misconceptions persist regarding the definition, incidence, severity and outcome of IC in clinical trials and the postmarketing setting. In this article, the frequency and clinical characteristics of IC associated with the use of alosetron and other serotonergic agents are examined, evidence of an association between IC and IBS is reviewed, and a scoring system to aid in the diagnosis of IC in any clinical situation is proposed.
Sigma-Aldrich
Alosetron hydrochloride, ≥98% (HPLC)