Merck
  • [Primary hemostasis activity in patients with arterial hypertension and impaired glucose tolerance treated with trandolapril].
[Primary hemostasis activity in patients with arterial hypertension and impaired glucose tolerance treated with trandolapril].
Klinicheskaia meditsina (2011-05-18)
V B Simonenko, I N Medvedev, O V Gamolina
ABSTRACT
The study was designed to evaluate effect of trandolapril, an ACE inhibitor, on platelet aggregation in patients with arterial hypertension and impaired glucose tolerance. The drug was given to 38 patients for 12 weeks. The key end points were dynamics of the blood lipid spectrum, lipid peroxidation in plasma and platelets, antioxidative protection of the blood's liquid medium and platelets, platelet aggregation. The data obtained were treated using Student's t-test. They show that trandolapril exerts positive effect on peroxidation syndrome and reduces platelet aggregation.
Sigma-Aldrich
Trandolapril, ≥98% (HPLC), white, powder