- Isolation, characterization, and production of red pigment from Cercospora piaropi a biocontrol agent for waterhyacinth.
Isolation, characterization, and production of red pigment from Cercospora piaropi a biocontrol agent for waterhyacinth.
Mycopathologia (2009-11-27)
Maricela Martínez Jiménez, Selenia Miranda Bahena, César Espinoza, Angel Trigos
PMID19941166
ABSTRACT
A red pigment produced by a Mexican isolate of Cercospora piaropi (waterhyacinth pathogen) has been isolated and identified as cercosporin. The kinetic of cercosporin production in culture media during dark/light regimes was evaluated. When C. piaropi was cultivated in continuous light and potato dextrose broth culture, a maximum of cercosporin production was observed (72.59 mg/l). Despite other reports, C piaropi Mexican isolate produce cercosporin in dark conditions (25.70 mg/l). The results suggest that production of cercosporin in C. piaropi-waterhyacinth pathogenesis is an important factor to take into account in biocontrol strategies.