Merck
  • An alternative DNA structure is necessary for pilin antigenic variation in Neisseria gonorrhoeae.
An alternative DNA structure is necessary for pilin antigenic variation in Neisseria gonorrhoeae.
Science (New York, N.Y.) (2009-08-08)
Laty A Cahoon, H Steven Seifert
ABSTRACT
Pathogens can use DNA recombination to promote antigenic variation (Av) of surface structures to avoid immune detection. We identified a cis-acting DNA sequence near the antigenically variable pilin locus of the human pathogen, Neisseria gonorrhoeae. This 16-base pair guanine (G)-rich sequence was required for pilin Av and formed a guanine quartet (G4) structure in vitro. Individual mutations that disrupted the structure also blocked pilin Av and prevented nicks required for recombination from occurring within the G4 region. A compound that binds and stabilizes G4 structures also inhibited pilin Av and prevented nicks from occurring on the G-rich strand. This site constitutes a recombination initiation sequence/structure that directs gene conversion to a specific chromosomal locus.
Sigma-Aldrich
Mesoporphyrin IX dihydrochloride, synthetic, 95%