- Allergic contact dermatitis from Laurus nobilis oil induced by massage.
Allergic contact dermatitis from Laurus nobilis oil induced by massage.
Contact dermatitis (2007-06-20)
Esra Adişen, Meltem Onder
PMID17577382
ABSTRACT
Non-occupational contact allergy to Laurus nobilis (laurel) oil is considered rare. Herein, we report a case of allergic contact dermatitis after a massage with a mixture of olive oil and L. nobilis oil. Patch testing showed (+++) reactions to laurel oil and (++) to the mixture of olive oil and laurel oil.