Merck
  • Chromatographic selectivity study of 4-fluorophenylacetic acid positional isomers separation.
Chromatographic selectivity study of 4-fluorophenylacetic acid positional isomers separation.
Journal of chromatography. A (2007-06-06)
Tyson Chasse, Robert Wenslow, Yuri Bereznitski
ABSTRACT
Unique properties of the fluorine atom stimulate widespread use and development of new organofluorine compounds in agrochemistry, biotechnology and pharmacology applications. However, relatively few synthetic methods exhibit a high degree of fluorination selectivity, which ultimately results in the presence of structurally related fluorinated isomers in the synthetic product. This outcome is undesirable from a pharmaceutical perspective as positional isomers possess different reactivity, biological activity and toxicity as compared to the desired product. It is advantageous to control positional isomers in the early stages of the synthetic process, as rejection and analysis of these isomers will likely become more difficult in later stages. The current work reports the development of a chromatographic analysis of 2- and 3-fluorophenylacetic acid positional isomer impurities in 4-fluorophenylacetic acid (4-FPAA), a building block in the synthesis of an active pharmaceutical ingredient. The method is employed as a part of a Quality by Design Approach to control purity of the starting material in order to eliminate the presence of undesirable positional isomers in the final drug substance. During method development, a wide range of chromatographic conditions and structurally related positional isomer probe molecules were exploited in an effort to gain insight into the specifics of the separation mechanism. For the systems studied it was shown that the choice of organic modifier played a key role in achieving acceptable separation. Further studies encompassed investigation of temperature influence on retention and selectivity of the FPAA isomers separation. Thermodynamic analysis of these data showed that the selectivity of the 2- and 4- fluorophenylacetic acids separation was dominated by an enthalpic process, while the selectivity of the 4- and 3-fluorophenylacetic acids separation was exclusively entropy driven (Delta(DeltaH degrees approximately 0). Studies of chromatographic behavior were complemented by solid state NMR experiments which provided valuable information regarding the relationship between stationary phase solvation and selectivity.
Sigma-Aldrich
4-Fluorophenylacetic acid, 98%