Merck
  • Mass spectrometric identification and quantification of 5-methoxytryptophol in quail retina.
Mass spectrometric identification and quantification of 5-methoxytryptophol in quail retina.
Biochemical and biophysical research communications (1989-12-29)
C W Tsang, S F Chan, P P Lee, S F Pang
ABSTRACT
The occurrence of 5-methoxytryptophol (5-MTL) in the quail retina was investigated by capillary column gas chromatography/mass spectrometry/selected ion monitoring using a deuterated internal standard. Based on ion intensity ratios in the mass spectra of pentafluoropropionyl and heptafluorobutyryl derivatives of 5-MTL and deuterated 5-MTL, 5-MTL was unequivocally identified in the quail retina. Similar to the circadian rhythm of retinal melatonin, retinal 5-MTL also exhibited a diurnal variation with high levels at mid-dark. However, no significant correlation between the diurnal levels of 5-MTL and melatonin was observed in the quail retina at mid-light or mid-dark.
Sigma-Aldrich
Sodium pentafluoropropionate, 98%
Sigma-Aldrich
Pentafluoropropionic acid, 97%