Merck
  • Preparation of succinimidyl and pentafluorophenyl active esters of 5- and 6-carboxyfluorescein.
Preparation of succinimidyl and pentafluorophenyl active esters of 5- and 6-carboxyfluorescein.
Bioconjugate chemistry (1997-03-01)
M Adamczyk, J R Fishpaugh, K J Heuser
ABSTRACT
A mixture of 5- and 6-carboxyfluorescein was activated with 1-[3-(dimethylamino)propyl]-3-ethylcarbodiimide hydrochloride in the presence of either N-hydroxysuccinimide or pentafluorophenol to give the corresponding succinimidyl and pentafluorophenyl esters. The regioisomeric mixtures were separated to give the 5- and 6-succinimidyl and pentafluorophenyl active esters in > 98% purity.
Sigma-Aldrich
Pentafluorophenol, ReagentPlus®, ≥99%