Merck
  • N-acetylputrescine as a characteristic constituent of cyanelle peptidoglycan in glaucocystophyte algae.
N-acetylputrescine as a characteristic constituent of cyanelle peptidoglycan in glaucocystophyte algae.
Journal of bacteriology (1996-12-01)
B Pfanzagl, G Allmaier, E R Schmid, M A de Pedro, W Löffelhardt
ABSTRACT
Cyanelle peptidoglycan from the glaucocystophyte algae Glaucocystis nostochinearum and Cyanoptyche gloeocystis was investigated by high-performance liquid chromatography of muropeptides, supported by matrix-assisted laser desorption-ionization mass spectrometry. The peptidoglycans of both species are modified with N-acetylputrescine, as has been demonstrated for cyanelle peptidoglycan of Cyanophora paradoxa.
Sigma-Aldrich
N-Acetylputrescine hydrochloride, ≥98% (TLC)