Merck
  • Optimum concentration and application time of 1,6-hexanediol as a dentin primer.
Optimum concentration and application time of 1,6-hexanediol as a dentin primer.
Dental materials journal (1995-12-01)
M A Rahman, C Tani, K Itoh, S Wakumoto, H Hisamitsu
ABSTRACT
The optimum concentration and application time for an experimental dentin primer composed of an aqueous solution of 1,6-hexanediol were determined by measuring the wall-to-wall polymerization contraction gap width of a commercial light-activated resin composite in a cylindrical dentin cavity prepared in an extracted human molar. Contraction gap formation was completely prevented only when the dentin cavity wall was primed with 45 wt% 1,6-hexanediol for 60 s. There were no significant differences in tensile bond strength among primers composed of 20.0-57.5 wt% 1,6-hexanediol. These results suggest that the optimum concentration and application time are 45 wt% and 60 s, respectively.
Sigma-Aldrich
1,6-Hexanediol, 99%
Sigma-Aldrich
1,6-Hexanediol, 97%
Sigma-Aldrich
1,6-Hexanediol solution, BioUltra, ~6 M in H2O