Merck
  • Analysis of Trimethoprim--sulfonamide drug combinations in dosage forms by UV spectroscopy and liquid chromatography (HPLC).
Analysis of Trimethoprim--sulfonamide drug combinations in dosage forms by UV spectroscopy and liquid chromatography (HPLC).
Farmaco (Societa chimica italiana : 1989) (1994-06-01)
D Bonazzi, V Andrisano, A M Di Pietra, V Cavrini
ABSTRACT
Derivative and difference spectrophotometric methods are described for the direct simultaneous analysis of combinations of Trimethoprim with sulfonamide drugs (sulfadiazine, sulfamethoxazole, sulfamethoxypyridazine) in commercial dosage forms. The more advantageous approach (derivative and difference mode) is suggested for each binary mixture. The assay results are compared with those obtained by a new chromatographic (HPLC) method, involving a C-18 column and a mobile phase (pH 6.5) containing 1.8 diaminooctane as amine modifier and sodium heptansulfonate as ion pairing agent.
Product Description
Supelco
Sulfamethoxypyridazine, VETRANAL®, analytical standard