Merck
  • Inhibition by antioxidants of agonist evoked cytosolic Ca++ increase, ATP secretion and aggregation of aspirinated human platelets.
Inhibition by antioxidants of agonist evoked cytosolic Ca++ increase, ATP secretion and aggregation of aspirinated human platelets.
Biochemical and biophysical research communications (1986-09-14)
A Alexandre, M G Doni, E Padoin, R Deana
ABSTRACT
The synthetic antioxidants butylated hydroxytoluene (BHT), nordihydroguaiaretic acid and the one-electron donor 1,1'-dimethylferrocene, inhibit cytosolic Ca++ increase, shape change, aggregation and ATP secretion in aspirinated washed human platelets stimulated by thrombin, vasopressin and platelet-activating factor. The antioxidants also inhibit cytosolic Ca++ increase originating from intracellular stores in the presence of EGTA. The effect of phorbol ester (TPA), which promotes platelet aggregation and secretion without raising the cytosolic Ca++, is also antioxidant-sensitive. Since agonist activation of aspirinated platelets does not involve cyclooxygenase or lipoxygenase metabolites, it is suggested that other yet unknown free radical-dependent pathways are involved in the mechanism of platelet activation, both in the protein kinase C-independent events leading to the cytosolic Ca++ increase, and in those, largely protein kinase C-dependent, leading to aggregation and ATP secretion.
Sigma-Aldrich
1,1′-Dimethylferrocene, 95%