Merck
  • Degradation and toxicity change of 4-chlorophenol in aqueous solution during CGDE treatment.
Degradation and toxicity change of 4-chlorophenol in aqueous solution during CGDE treatment.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-05-17)
Xing-long Jin, Xiao-qing Zhao, Xiao-yan Wang, Zhi-rong Wang
ABSTRACT
This study focused on the degradation and toxicity change of 4-chlorophenol (4-CP) in aqueous solution by contact glow discharge electrolysis (CGDE) with and without Fe(2+). It was found that the addition of Fe(2+) increased the degradation rate of 4-CP and decreased the toxicity of the solution. After 90 min CGDE treatment, only 28.0% 4-CP was removed and EC50 increased from 33.8 to 63.1 mg/L. With 25.0 mg/L Fe(2+) in solution, the degradation rate of 4-CP reached 93.3% and EC50 changed from 35.9 to 93.9 mg/L after 90 min CGDE treatment.
Sigma-Aldrich
4-Chlorophenol, ≥99%