Merck
  • Prospective study of histomorphometry, biochemical bone markers and bone densitometric response to pamidronate in β-thalassaemia presenting with osteopenia-osteoporosis syndrome.
Prospective study of histomorphometry, biochemical bone markers and bone densitometric response to pamidronate in β-thalassaemia presenting with osteopenia-osteoporosis syndrome.
British journal of haematology (2012-09-13)
Ratna Chatterjee, Farrukh T Shah, Bernard A Davis, Margaret Byers, Dev Sooranna, Rekha Bajoria, Jean Pringle, John B Porter
ABSTRACT
This study aimed to evaluate bone remodelling disorders in thalassaemia by using pamidronate (PD) infusion with or without hormone replacement therapy (HRT) as a diagnostic-therapeutic tool. In this prospective study, 24 adult thalassaemia major (TM) and 10 thalassaemia intermedia (TI) patients received either PD and HRT or HRT only (controls) for 3 years. Eugonadal patients with TI had PD only. Bone remodelling was assessed by dual energy X ray absorptiometry (DXA scan), type 1-collagen biochemical bone markers (BBM) and histomorphometry of iliac crest biopsy before and after PD. As a group, thalassaemics had a significant improvement in spinal and femoral bone mineral density Z scores following PD (P < 0·01) compared to the controls. Although BBM were comparable pre-therapy, they were significantly lower in the PD cohort (P < 0·001) compared to the control group. All patients had osteopenia, diminished osteoid formation and bone volume on histomorphometry pre-therapy with high turnover bone disease (HTO) in TM and low-turnover disease (LTO) in TI. In TM, bone volume improved significantly, whereas TI patients showed little or no response to PD. In conclusion, histomorphometry data suggest that TM patients have a distinct pathology of high turnover bone disease compared to TI patients, who have low-turnover disease.
Sigma-Aldrich
Pamidronate disodium salt hydrate, ≥95% (NMR), solid