Merck
  • Chiral Brønsted acid catalyzed enantioselective allenylation of aldehydes.
Chiral Brønsted acid catalyzed enantioselective allenylation of aldehydes.
Chemical communications (Cambridge, England) (2012-08-09)
Leleti Rajender Reddy
ABSTRACT
A versatile and highly enantioselective chiral Brønsted acid-catalyzed allenylation of aldehydes with propargyl borolane is reported. The reaction is shown to be practical and quite general with a broad substrate scope covering aryl, heteroaryl, α,β-unsaturated, and aliphatic aldehydes.
Sigma-Aldrich
Pargyline hydrochloride
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%