Merck
  • Design, synthesis, and biological activities of some branched carbasugars: construction of a substituted 6-oxabicyclo[3.2.1]nonane skeleton.
Design, synthesis, and biological activities of some branched carbasugars: construction of a substituted 6-oxabicyclo[3.2.1]nonane skeleton.
The Journal of organic chemistry (2012-05-23)
Arif Baran, Sinem Çambul, Mehmet Nebioglu, Metin Balci
ABSTRACT
Transformation of cyclohexa-2,4-diene-1,2-diylbis(methylene) diacetate to various carbasugars is described. Photooxygenation of a cyclohexadiene derivative gave a bicyclicendoperoxide, which was reduced with thiourea to [2-[(acetyloxy)methyl]cyclohexa-2,4-dien-1-yl]methyl acetate. Epoxidation of the remaining double bond followed by epoxide ring-opening and hydrolysis of the acetate groups gave one of the target hexols. The bicyclic endoperoxide was rearranged to a diepoxide with CoTPP. The diepoxide was reacted with sulfamic acid in acetic anhydride, resulting in the formation of a new branched carbasugar as well as in the formation of cyclitols with a 6-oxabicyclo[3.2.1]nonane skeleton. The mechanism of the formation of the products is discussed. The inhibition activity of six cyclitol derivatives was tested against α-glycosidase.
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Nonane, ReagentPlus®, 99%
Supelco
Nonane, analytical standard