Merck
  • Poly(ADP-ribose) polymerase inhibition improves erectile function by activation of nitric oxide/cyclic guanosine monophosphate pathway in diabetic rats.
Poly(ADP-ribose) polymerase inhibition improves erectile function by activation of nitric oxide/cyclic guanosine monophosphate pathway in diabetic rats.
The journal of sexual medicine (2012-03-21)
Wen Ji Li, YuBing Peng, Juan Zhou, Bao Li, Hao Wang, Jun Zhang, Zhong Wang
ABSTRACT
Endothelial dysfunction-induced abnormalities of the nitric oxide (NO)/cyclic guanosine monophosphate (cGMP) signaling pathway in the corpus cavernosum are thought to be the main factors involved in the pathogenesis of diabetes-induced erectile dysfunction (ED). Recent studies have shown that the poly(adenosine diphosphate ribose) polymerase (PARP) pathway plays a critical role in diabetic endothelial dysfunction. The aim of this study is to determine whether activation of the PARP pathway is involved in diabetic cavernosal endothelial dysfunction and abnormalities of the NO/cGMP pathway. Male Sprague-Dawley rats were randomly divided into three groups: age-matched controls, diabetic controls (DM), and the 3-aminobenzamide (3-AB, a PARP inhibitor)-treated diabetic group (DM+3-AB). Diabetes was induced by intraperitoneal injection of streptozotocin. Eight weeks after inducing diabetes, the DM+3-AB group was treated with 3-AB for 4 weeks. Erectile function was assessed at 12 weeks after inducing diabetes by stimulating the cavernous nerve. Expression of poly(ADP-ribose), protein kinase B (Akt), phospho-Akt, endothelial nitric oxide synthase (eNOS), phospho-eNOS, and neuronal nitric oxide synthase (nNOS) were evaluated by Western blot. Cavernous NO generation and cGMP levels were also determined. The DM group showed impaired erectile function and significantly increased PARP activity. Expression of total eNOS and nNOS, phospho-Akt, and eNOS decreased significantly in the DM group compared with those in the control group. In addition, cavernous NO generation and cGMP levels decreased significantly in the DM group compared with those in the control group. Treatment with 3-AB restored erectile function and significantly reversed all molecular alterations except decreased nNOS expression. Overactivation of the PARP pathway in the corpus cavernosum of diabetic rats was involved in cavernosal endothelial dysfunction and abnormalities of the NO/cGMP pathway resulting in ED. These findings may be applied to develop novel therapies for patients with diabetic ED.
Sigma-Aldrich
3-Aminobenzamide, ≥99% (TLC)