- [Isolation of the bronchi with intraoperative packing in children with bronchiectasis].
[Isolation of the bronchi with intraoperative packing in children with bronchiectasis].
Grudnaia i serdechno-sosudistaia khirurgiia (1990-01-01)
E A Stepanov, L A Sitko, E S Bocharnikov, S A Schastnyĭ
PMID2192747
ABSTRACT
The authors discuss experience in 168 operations for extirpation and segmental resection of affected bronchi in children with bronchiectases. To prevent recanalization of the bronchus after its segmental resection and to prevent the formation of an abscess in the isolated part of the lung they suggest a method for intraoperative packing of the remaining bronchi with biopolymer material.
Sigma-Aldrich
Poly(propylene glycol), tolylene 2,4-diisocyanate terminated, average Mn ~2,300 (narrow MW distribution), isocyanate ~3.6 wt. %