Merck
  • A rapid sample screening method for authenticity control of whiskey using capillary electrophoresis with online preconcentration.
A rapid sample screening method for authenticity control of whiskey using capillary electrophoresis with online preconcentration.
Journal of agricultural and food chemistry (2011-06-15)
Melina Heller, Luciano Vitali, Marcone Augusto Leal Oliveira, Ana Carolina O Costa, Gustavo Amadeu Micke
ABSTRACT
The present study aimed to develop a methodology using capillary electrophoresis for the determination of sinapaldehyde, syringaldehyde, coniferaldehyde, and vanillin in whiskey samples. The main objective was to obtain a screening method to differentiate authentic samples from seized samples suspected of being false using the phenolic aldehydes as chemical markers. The optimized background electrolyte was composed of 20 mmol L(-1) sodium tetraborate with 10% MeOH at pH 9.3. The study examined two kinds of sample stacking, using a long-end injection mode: normal sample stacking (NSM) and sample stacking with matrix removal (SWMR). In SWMR, the optimized injection time of the samples was 42 s (SWMR42); at this time, no matrix effects were observed. Values of r were >0.99 for the both methods. The LOD and LOQ were better than 100 and 330 mg mL(-1) for NSM and better than 22 and 73 mg L(-1) for SWMR. The CE-UV reliability in the aldehyde analysis in the real sample was compared statistically with LC-MS/MS methodology, and no significant differences were found, with a 95% confidence interval between the methodologies.
Sigma-Aldrich
Syringaldehyde, 98%
Sigma-Aldrich
Syringaldehyde, ≥98%, FG