Merck
  • Macromolecular design of aliphatic polyesters with maintained mechanical properties and a rapid, customized degradation profile.
Macromolecular design of aliphatic polyesters with maintained mechanical properties and a rapid, customized degradation profile.
Biomacromolecules (2011-05-03)
Sofia Malberg, Anders Hoglund, Ann-Christine Albertsson
ABSTRACT
An innovative type of triblock copolymer that maintains and even increases the mechanical properties of poly(l-lactide) (PLLA) and poly(ε-caprolactone) (PCL) with a controlled, predictable, and rapid degradation profile has been synthesized. Elastic triblock copolymers were formed from the hydrophobic and crystalline PLLA and PCL with an amorphous and hydrophilic middle block of poly(but-2-ene-1,4-diyl malonate) (PBM). The polymers were subjected to degradation in PBS at 37 °C for up to 91 days. Prior to degradation, ductility of the PLLA-PBM-PLLA was approximately 4 times greater than that of the homopolymer of PLLA, whereas the modulus and tensile stress at break were unchanged. A rapid initial hydrolysis in the amorphous PBM middle block changed the microstructure from triblock to diblock with a significant reduction in ductility and molecular weight. The macromolecular structure of the triblock copolymer of PLLA and PBM generates a more flexible and easier material to handle during implant, with the advantage of a customized degradation profile, demonstrating its potential use in future biomedical applications.
Sigma-Aldrich
Polybutenes, average Mn ~920 by VPO, isobutylene >90 %
Sigma-Aldrich
Polybutenes, average Mn ~2,300 by VPO, isobutylene >90 %